Tisk…

8-Bromo-4-chloro-5-fluoroquinoline 95%

Dodavatel: Apollo Scientific
Varování

PC499015-1G PC499015-250MG
APOSPC499015-1GEA 8700 CZK
APOSPC499015-1G APOSPC499015-250MG
8-Bromo-4-chloro-5-fluoroquinoline 95%
8-Bromo-4-chloro-5-fluoroquinoline
Vzorec: C₉H₄BrClFN
MW: 260,49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527289
CAS číslo: 1065093-11-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR