6-Bromo-8-fluoro-4-hydroxyquinoline 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC499014-1G
PC499014-250MG
APOSPC499014-1GEA
14490
CZK
APOSPC499014-1G
APOSPC499014-250MG
6-Bromo-8-fluoro-4-hydroxyquinoline 95%
6-Bromo-8-fluoro-4-hydroxyquinoline
Vzorec:
C₉H₅BrFNO MW: 242,045 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09861332 CAS číslo: 1019016-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...