4,6-Dichloro-8-fluoroquinoline 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC499013-1G
PC499013-250MG
APOSPC499013-1GEA
24770
CZK
APOSPC499013-1G
APOSPC499013-250MG
4,6-Dichloro-8-fluoroquinoline 95%
4,6-Dichloro-8-fluoroquinoline
Vzorec:
C₉H₄Cl₂FN MW: 216,039 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09861349 CAS číslo: 1019016-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...