2-Fluoro-4-methoxybenzotrifluoride
Dodavatel: Apollo Scientific
PC499008-1G
PC499008-5G
APOSPC499008-1GEA
6250
CZK
APOSPC499008-1G
APOSPC499008-5G
2-Fluoro-4-methoxybenzotrifluoride
2-Fluoro-4-methoxybenzotrifluoride
Vzorec:
C₈H₆F₄O MW: 194,126 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13185641 CAS číslo: 1214344-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...