Tisk…

2’-Bromo-5’-(trifluoromethoxy)acetophenone 95%

Dodavatel: Apollo Scientific

Synonyms: 1-[2-Bromo-5-(trifluoromethoxy)phenyl]ethanone

PC49788-1G PC49788-250MG PC49788-5G
APOSPC49788-1GEA 12200 CZK
APOSPC49788-1G APOSPC49788-250MG APOSPC49788-5G
2’-Bromo-5’-(trifluoromethoxy)acetophenone 95%
2’-Bromo-5’-(trifluoromethoxy)acetophenone
Vzorec: C₉H₆BrF₃O₂
MW: 283,042 g/mol
Storage Temperature: Ambient
CAS číslo: 1558247-47-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR