Tisk…

Bis(trifluoromethanesulphonyl)methane 95%

Dodavatel: Apollo Scientific

Synonyms: Bis((trifluoromethyl)sulphonyl)methane, Bis(trifluoromethylsulphonyl)methane

PC49716-250MG PC49716-5G
APOSPC49716-250MGEA 0 CZK
APOSPC49716-250MG APOSPC49716-5G
Bis(trifluoromethanesulphonyl)methane 95%
Bis(trifluoromethanesulphonyl)methane
Vzorec: C₃H₂F₆O₄S₂
MW: 280.166 g/mol
Bod varu: 191 °C (745 mm Hg)
Bod tání: 32…36 °C
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01861122
CAS číslo: 428-76-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR