2-Bromo-4-(difluoromethyl)-1-iodobenzene 95%
Dodavatel: Apollo Scientific
PC49705-1G
PC49705-250MG
APOSPC49705-1GEA
16340
CZK
APOSPC49705-1G
APOSPC49705-250MG
2-Bromo-4-(difluoromethyl)-1-iodobenzene 95%
2-Bromo-4-(difluoromethyl)-1-iodobenzene
Vzorec:
C₇H₄BrF₂I Storage Temperature: Ambient |
MDL Number:
MFCD18391106 CAS číslo: 1261642-33-4 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...