Tisk…

4-Fluoro-1H-pyrrolo[2,3-b]pyridine-5-carbonitrile 97%

Dodavatel: Apollo Scientific
Varování

PC49682-100MG PC49682-250MG PC49682-50MG
APOSPC49682-100MGEA 5590 CZK
APOSPC49682-100MG APOSPC49682-250MG APOSPC49682-50MG
4-Fluoro-1H-pyrrolo[2,3-b]pyridine-5-carbonitrile 97%
4-Fluoro-1H-pyrrolo[2,3-b]pyridine-5-carbonitrile
Vzorec: C₈H₄FN₃
Storage Temperature: Ambient
MDL Number: MFCD18380908
CAS číslo: 1260381-44-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR