Tisk…

Methyl 2,3-difluoro-5-methylbenzoate

Dodavatel: Apollo Scientific
Varování

PC49668-1G PC49668-250MG
APOSPC49668-1GEA 16370 CZK
APOSPC49668-1G APOSPC49668-250MG
Methyl 2,3-difluoro-5-methylbenzoate
Methyl 2,3-difluoro-5-methylbenzoate
Vzorec: C₉H₈F₂O₂
MW: 186,155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042838
CAS číslo: 1314919-70-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR