Manganese bis(trifluoromethanesulphonate)
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC49662-1G
PC49662-25G
PC49662-5G
APOSPC49662-1GEA
744
CZK
APOSPC49662-1G
APOSPC49662-25G
APOSPC49662-5G
Manganese bis(trifluoromethanesulphonate)
Manganese bis(trifluoromethanesulphonate)
Vzorec:
2(CF₃O₃S)·Mn MW: 353.076 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17015267 CAS číslo: 55120-76-8 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...