Tisk…

4,4,5,5,5-Pentafluoropentyl methanesulphonate

Dodavatel: Apollo Scientific
Varování

PC49661-1G PC49661-5G
APOSPC49661-1GEA 7080 CZK
APOSPC49661-1G APOSPC49661-5G
4,4,5,5,5-Pentafluoropentyl methanesulphonate
4,4,5,5,5-Pentafluoropentyl methanesulphonate
Vzorec: C₆H₉F₅O₃S
Storage Temperature: Ambient
MDL Number: MFCD04972956
CAS číslo: 252947-01-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR