Methyl 2-fluoro-5-hydroxybenzoate
Dodavatel: Apollo Scientific
|
Varování
|
PC49647-10G
PC49647-1G
PC49647-5G
APOSPC49647-10GEA
6420
CZK
APOSPC49647-10G
APOSPC49647-1G
APOSPC49647-5G
Methyl 2-fluoro-5-hydroxybenzoate
Methyl 2-fluoro-5-hydroxybenzoate
Vzorec:
C₈H₇FO₃ MW: 170.138 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12406843 CAS číslo: 1084801-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...