5-Iodo-2-methoxy-3-(trifluoromethyl)benzonitrile 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
5-Iodo-3-(trifluoromethyl)-o-anisonitrile, 3-Cyano-5-iodo-2-methoxybenzotrifluoride
PC49623-1G
PC49623-250MG
APOSPC49623-1GEA
4960
CZK
APOSPC49623-1G
APOSPC49623-250MG
5-Iodo-2-methoxy-3-(trifluoromethyl)benzonitrile 97%
5-Iodo-2-methoxy-3-(trifluoromethyl)benzonitrile
Vzorec:
C₉H₅F₃INO MW: 327,042 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD25541890 CAS číslo: 1445995-74-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...