Tisk…

5-Bromo-2-hydroxy-3-(trifluoromethyl)benzoic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-3-carboxy-2-hydroxybenzotrifluoride, 5-Bromo-3-(trifluoromethyl)salicylic acid

PC49615-1G PC49615-250MG
APOSPC49615-1GEA 6620 CZK
APOSPC49615-1G APOSPC49615-250MG
5-Bromo-2-hydroxy-3-(trifluoromethyl)benzoic acid 97%
5-Bromo-2-hydroxy-3-(trifluoromethyl)benzoic acid
Vzorec: C₈H₄BrF₃O₃
MW: 285,015 g/mol
Bod tání: 185…188 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06203616
CAS číslo: 251300-31-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR