Tisk…

2-Hydroxy-5-iodo-3-(trifluoromethyl)benzoic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Carboxy-2-hydroxy-5-iodobenzotrifluoride, 5-Iodo-3-(trifluoromethyl)salicylic acid

PC49614-1G PC49614-250MG
APOSPC49614-1GEA 3970 CZK
APOSPC49614-1G APOSPC49614-250MG
2-Hydroxy-5-iodo-3-(trifluoromethyl)benzoic acid 97%
2-Hydroxy-5-iodo-3-(trifluoromethyl)benzoic acid
Vzorec: C₈H₄F₃IO₃
MW: 332.015 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD25541889
CAS číslo: 1445995-72-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR