Tisk…

2-Iodo-6-nitro-4-[(trifluoromethyl)sulphonyl]phenol

Dodavatel: Apollo Scientific
Varování

PC49600-1G PC49600-250MG
APOSPC49600-1GEA 7250 CZK
APOSPC49600-1G APOSPC49600-250MG
2-Iodo-6-nitro-4-[(trifluoromethyl)sulphonyl]phenol
2-Iodo-6-nitro-4-[(trifluoromethyl)sulphonyl]phenol
Vzorec: C₇H₃F₃INO₅S
Storage Temperature: Refrigerator
MDL Number: MFCD24850031
CAS číslo: 1440535-20-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR