5-Ethynyl-2-fluorobenzaldehyde 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Fluoro-3-formylphenylacetylene
PC49599-1G
PC49599-250MG
APOSPC49599-1GEA
7050
CZK
APOSPC49599-1G
APOSPC49599-250MG
5-Ethynyl-2-fluorobenzaldehyde 97%
5-Ethynyl-2-fluorobenzaldehyde
Vzorec:
C₉H₅FO MW: 148.134 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD24850036 CAS číslo: 1440535-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...