Tisk…

Iron(II) tetrafluoroborate hexahydrate 97%

Dodavatel: Apollo Scientific

Synonyms: Ferrous tetrafluoroborate

PC49590-100G PC49590-25G
APOSPC49590-100GEA 13890 CZK
APOSPC49590-100G APOSPC49590-25G
Iron(II) tetrafluoroborate hexahydrate 97%
Iron(II) tetrafluoroborate hexahydrate
Vzorec: 2(BF₄)·Fe·6H₂O
MW: 337,546 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00192235
CAS číslo: 13877-16-2
UN: 3260
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR