Methyl 6-bromo-4-chloro-2,3-difluorobenzoate 95%
Dodavatel: Apollo Scientific
PC49468-1G
PC49468-250MG
PC49468-5G
APOSPC49468-1GEA
14880
CZK
APOSPC49468-1G
APOSPC49468-250MG
APOSPC49468-5G
Methyl 6-bromo-4-chloro-2,3-difluorobenzoate 95%
Methyl 6-bromo-4-chloro-2,3-difluorobenzoate
Vzorec:
C₈H₄BrClF₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD32662396 CAS číslo: 2414146-32-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...