Tisk…

5-(Difluoromethoxy)-2-fluoropyridine 95%

Dodavatel: Apollo Scientific
Varování

PC49467-1G PC49467-250MG PC49467-5G
APOSPC49467-1GEA 15640 CZK
APOSPC49467-1G APOSPC49467-250MG APOSPC49467-5G
5-(Difluoromethoxy)-2-fluoropyridine 95%
5-(Difluoromethoxy)-2-fluoropyridine
Vzorec: C₆H₄F₃NO
Storage Temperature: Ambient
MDL Number: MFCD13185839
CAS číslo: 130318-80-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR