Tisk…

3,5-Difluoro-2-nitrophenylacetic acid

Dodavatel: Apollo Scientific
Varování

PC49466-1G PC49466-250MG
APOSPC49466-1GEA 14880 CZK
APOSPC49466-1G APOSPC49466-250MG
3,5-Difluoro-2-nitrophenylacetic acid
3,5-Difluoro-2-nitrophenylacetic acid
Vzorec: C₈H₅F₂NO₄
MW: 217,126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26801851
CAS číslo: 130400-45-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR