4-Nitro-2-(trifluoromethyl)phenylacetic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC49465-1G
PC49465-250MG
PC49465-5G
APOSPC49465-1GEA
6420
CZK
APOSPC49465-1G
APOSPC49465-250MG
APOSPC49465-5G
4-Nitro-2-(trifluoromethyl)phenylacetic acid
4-Nitro-2-(trifluoromethyl)phenylacetic acid
Vzorec:
C₉H₆F₃NO₄ MW: 249,143 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11036389 CAS číslo: 199469-91-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...