Tisk…

5-Bromo-3-fluoro-2-nitrophenylacetic acid

Dodavatel: Apollo Scientific
Varování

PC49464-1G PC49464-250MG PC49464-5G
APOSPC49464-1GEA 4960 CZK
APOSPC49464-1G APOSPC49464-250MG APOSPC49464-5G
5-Bromo-3-fluoro-2-nitrophenylacetic acid
5-Bromo-3-fluoro-2-nitrophenylacetic acid
Vzorec: C₈H₅BrFNO₄
Storage Temperature: Ambient
MDL Number: MFCD28740371
CAS číslo: 1805938-07-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR