Tisk…

6-Bromo-4-chloro-2,3-difluorobenzaldehyde 95%

Dodavatel: Apollo Scientific

PC49462-1G PC49462-250MG
APOSPC49462-1GEA 11540 CZK
APOSPC49462-1G APOSPC49462-250MG
6-Bromo-4-chloro-2,3-difluorobenzaldehyde 95%
6-Bromo-4-chloro-2,3-difluorobenzaldehyde
Vzorec: C₇H₂BrClF₂O
MW: 255,444 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32662403
CAS číslo: 2514941-95-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR