Tisk…

6-Bromo-4-chloro-2,3-difluorophenol 95%

Dodavatel: Apollo Scientific

PC49461-1G PC49461-250MG PC49461-5G
APOSPC49461-1GEA 12960 CZK
APOSPC49461-1G APOSPC49461-250MG APOSPC49461-5G
6-Bromo-4-chloro-2,3-difluorophenol 95%
6-Bromo-4-chloro-2,3-difluorophenol
Vzorec: C₆H₂BrClF₂O
MW: 243.433 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32662402
CAS číslo: 1935997-11-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR