Tisk…

2-Bromo-4,6-difluorobenzaldehyde 95%

Dodavatel: Apollo Scientific
Varování

PC49452-1G PC49452-250MG PC49452-5G
APOSPC49452-1GEA 8070 CZK
APOSPC49452-1G APOSPC49452-250MG APOSPC49452-5G
2-Bromo-4,6-difluorobenzaldehyde 95%
2-Bromo-4,6-difluorobenzaldehyde
Vzorec: C₇H₃BrF₂O
MW: 220,999 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18642940
CAS číslo: 154650-59-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR