Tisk…

2-Fluoro-4-methyl-5-nitrobenzoyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

PC49449-1G PC49449-250MG
APOSPC49449-1GEA 25130 CZK
APOSPC49449-1G APOSPC49449-250MG
2-Fluoro-4-methyl-5-nitrobenzoyl chloride
2-Fluoro-4-methyl-5-nitrobenzoyl chloride
Vzorec: C₈H₅ClFNO₃
Storage Temperature: Ambient
MDL Number: MFCD28797714
CAS číslo: 1806427-38-2
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR