Methyl 3-(chlorosulphonyl)-4-fluorobenzoate
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC49447-1G
PC49447-250MG
PC49447-5G
APOSPC49447-1GEA
9530
CZK
APOSPC49447-1G
APOSPC49447-250MG
APOSPC49447-5G
Methyl 3-(chlorosulphonyl)-4-fluorobenzoate
Methyl 3-(chlorosulphonyl)-4-fluorobenzoate
Vzorec:
C₈H₆ClFO₄S Storage Temperature: Ambient |
MDL Number:
MFCD11650667 CAS číslo: 1099660-67-9 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...