2-Bromo-6-chloro-4-fluorobenzaldehyde 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC49437-1G
PC49437-5G
PC49437-250MG
APOSPC49437-1GEA
3940
CZK
APOSPC49437-1G
APOSPC49437-5G
APOSPC49437-250MG
2-Bromo-6-chloro-4-fluorobenzaldehyde 95%
2-Bromo-6-chloro-4-fluorobenzaldehyde
Vzorec:
C₇H₃BrClFO MW: 237,454 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28733865 CAS číslo: 1433990-64-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...