Tisk…

2-Bromo-4-fluoro-6-(trifluoromethyl)benzoic acid 95%

Dodavatel: Apollo Scientific
Varování

PC49433-1G PC49433-250MG
APOSPC49433-1GEA 1660 CZK
APOSPC49433-1G APOSPC49433-250MG
2-Bromo-4-fluoro-6-(trifluoromethyl)benzoic acid 95%
2-Bromo-4-fluoro-6-(trifluoromethyl)benzoic acid
Vzorec: C₈H₃BrF₄O₂
MW: 287.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31759995
CAS číslo: 2091606-46-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR