Tisk…

2-Bromo-6-fluoro-4-(trifluoromethyl)benzoic acid 95%

Dodavatel: Apollo Scientific
Varování

PC49432-1G PC49432-250MG
APOSPC49432-1GEA 12870 CZK
APOSPC49432-1G APOSPC49432-250MG
2-Bromo-6-fluoro-4-(trifluoromethyl)benzoic acid 95%
2-Bromo-6-fluoro-4-(trifluoromethyl)benzoic acid
Vzorec: C₈H₃BrF₄O₂
Storage Temperature: Ambient
MDL Number: MFCD31759997
CAS číslo: 1800423-60-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR