3-Bromo-2-fluoro-5-methylbenzoic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC49431-1G
PC49431-25G
PC49431-5G
PC49431-10G
APOSPC49431-1GEA
910
CZK
APOSPC49431-1G
APOSPC49431-25G
APOSPC49431-5G
APOSPC49431-10G
3-Bromo-2-fluoro-5-methylbenzoic acid 95%
3-Bromo-2-fluoro-5-methylbenzoic acid
Vzorec:
C₈H₆BrFO₂ MW: 233,034 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00506954 CAS číslo: 72518-16-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...