Tisk…

2-Cyclopropyl-5-fluoroaniline

Dodavatel: Apollo Scientific
Varování

PC49423-1G PC49423-250MG PC49423-5G
APOSPC49423-1GEA 2200 CZK
APOSPC49423-1G APOSPC49423-250MG APOSPC49423-5G
2-Cyclopropyl-5-fluoroaniline
2-Cyclopropyl-5-fluoroaniline
Vzorec: C₉H₁₀FN
MW: 151,181 g/mol
Storage Temperature: Freezer
MDL Number: MFCD30322597
CAS číslo: 1538364-89-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR