2-Cyclopropyl-3-fluoroaniline
Dodavatel: Apollo Scientific
|
Varování
|
PC49419-1G
PC49419-250MG
PC49419-5G
APOSPC49419-1GEA
2480
CZK
APOSPC49419-1G
APOSPC49419-250MG
APOSPC49419-5G
2-Cyclopropyl-3-fluoroaniline
2-Cyclopropyl-3-fluoroaniline
Vzorec:
C₉H₁₀FN Storage Temperature: Freezer |
MDL Number:
MFCD31793572 CAS číslo: 2366994-64-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...