3-Cyclopropyl-2-fluoroaniline
Dodavatel: Apollo Scientific
|
Varování
|
PC49415-1G
PC49415-250MG
PC49415-5G
APOSPC49415-1GEA
2280
CZK
APOSPC49415-1G
APOSPC49415-250MG
APOSPC49415-5G
3-Cyclopropyl-2-fluoroaniline
3-Cyclopropyl-2-fluoroaniline
Vzorec:
C₉H₁₀FN Storage Temperature: Freezer |
MDL Number:
MFCD23163094 CAS číslo: 1386459-75-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...