Tisk…

1-Cyclopropyl-4-fluoro-2-nitrobenzene

Dodavatel: Apollo Scientific
Varování

PC49414-1G PC49414-250MG PC49414-5G
APOSPC49414-1GEA 12340 CZK
APOSPC49414-1G APOSPC49414-250MG APOSPC49414-5G
1-Cyclopropyl-4-fluoro-2-nitrobenzene
1-Cyclopropyl-4-fluoro-2-nitrobenzene
Vzorec: C₉H₈FNO₂
MW: 181,164 g/mol
Storage Temperature: Freezer
MDL Number: MFCD31793570
CAS číslo: 2366994-53-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR