2-Cyclopropyl-1-fluoro-4-nitrobenzene
Dodavatel: Apollo Scientific
|
Varování
|
PC49413-1G
PC49413-250MG
PC49413-5G
APOSPC49413-1GEA
13560
CZK
APOSPC49413-1G
APOSPC49413-250MG
APOSPC49413-5G
2-Cyclopropyl-1-fluoro-4-nitrobenzene
2-Cyclopropyl-1-fluoro-4-nitrobenzene
Vzorec:
C₉H₈FNO₂ Storage Temperature: Freezer |
MDL Number:
MFCD31793569 CAS číslo: 2366994-57-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...