1-Cyclopropyl-3-fluoro-5-nitrobenzene
Dodavatel: Apollo Scientific
|
Varování
|
PC49411-1G
PC49411-250MG
PC49411-5G
APOSPC49411-1GEA
1120
CZK
APOSPC49411-1G
APOSPC49411-250MG
APOSPC49411-5G
1-Cyclopropyl-3-fluoro-5-nitrobenzene
1-Cyclopropyl-3-fluoro-5-nitrobenzene
Vzorec:
C₉H₈FNO₂ MW: 181,164 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD31793567 CAS číslo: 2366994-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...