1-Cyclopropyl-2-fluoro-4-nitrobenzene
Dodavatel: Apollo Scientific
|
Varování
|
PC49410-1G
PC49410-250MG
PC49410-5G
APOSPC49410-1GEA
910
CZK
APOSPC49410-1G
APOSPC49410-250MG
APOSPC49410-5G
1-Cyclopropyl-2-fluoro-4-nitrobenzene
1-Cyclopropyl-2-fluoro-4-nitrobenzene
Vzorec:
C₉H₈FNO₂ MW: 181,164 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD31793566 CAS číslo: 2366994-63-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...