Tisk…

4-Cyclopropyl-1-fluoro-2-nitrobenzene

Dodavatel: Apollo Scientific
Varování

PC49407-1G PC49407-250MG PC49407-5G
APOSPC49407-1GEA 12400 CZK
APOSPC49407-1G APOSPC49407-250MG APOSPC49407-5G
4-Cyclopropyl-1-fluoro-2-nitrobenzene
4-Cyclopropyl-1-fluoro-2-nitrobenzene
Vzorec: C₉H₈FNO₂
MW: 181,164 g/mol
Storage Temperature: Freezer
MDL Number: MFCD17677330
CAS číslo: 769159-85-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR