Tisk…

4-Cyclopropyl-2-fluoro-1-nitrobenzene

Dodavatel: Apollo Scientific
Varování

PC49406-1G PC49406-250MG PC49406-5G
APOSPC49406-1GEA 992 CZK
APOSPC49406-1G APOSPC49406-250MG APOSPC49406-5G
4-Cyclopropyl-2-fluoro-1-nitrobenzene
4-Cyclopropyl-2-fluoro-1-nitrobenzene
Vzorec: C₉H₈FNO₂
MW: 181,164 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22418246
CAS číslo: 932700-92-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR