1-Cyclopropyl-2-fluoro-3-nitrobenzene
Dodavatel: Apollo Scientific
|
Varování
|
PC49405-1G
PC49405-250MG
PC49405-5G
APOSPC49405-1GEA
744
CZK
APOSPC49405-1G
APOSPC49405-250MG
APOSPC49405-5G
1-Cyclopropyl-2-fluoro-3-nitrobenzene
1-Cyclopropyl-2-fluoro-3-nitrobenzene
Vzorec:
C₉H₈FNO₂ MW: 181,164 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD31793563 CAS číslo: 2366994-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...