Tisk…

Ethyl 8-fluoroimidazo[1,2-a]pyridine-2-carboxylate

Dodavatel: Apollo Scientific
Varování

PC49394-1G PC49394-250MG
APOSPC49394-1GEA 13560 CZK
APOSPC49394-1G APOSPC49394-250MG
Ethyl 8-fluoroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 8-fluoroimidazo[1,2-a]pyridine-2-carboxylate
Vzorec: C₁₀H₉FN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD13195311
CAS číslo: 1260843-88-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR