Tisk…

S-(2-Amino-4-fluorophenyl) dimethylcarbamothioate 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-[(Dimethylcarbamoyl)sulphanyl]-5-fluoroaniline, S-(2-Amino-4-fluorophenyl) dimethylthiocarbamate

PC49381-1G PC49381-250MG
APOSPC49381-1GEA 15870 CZK
APOSPC49381-1G APOSPC49381-250MG
S-(2-Amino-4-fluorophenyl) dimethylcarbamothioate 97%
S-(2-Amino-4-fluorophenyl) dimethylcarbamothioate
Vzorec: C₉H₁₁FN₂OS
MW: 214,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24636943
CAS číslo: 1820674-43-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR