Tisk…

4-Fluoro-3-iodobenzyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: α-Bromo-4-fluoro-3-iodotoluene, 4-(Bromomethyl)-1-fluoro-2-iodobenzene

PC49377-1G PC49377-250MG
APOSPC49377-1GEA 7440 CZK
APOSPC49377-1G APOSPC49377-250MG
4-Fluoro-3-iodobenzyl bromide 97%
4-Fluoro-3-iodobenzyl bromide
Vzorec: C₇H₅BrFI
MW: 314,921 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08236849
CAS číslo: 260050-97-3
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR