2-Fluoro-3-iodobenzaldehyde 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC49373-1G
PC49373-5G
APOSPC49373-1GEA
2650
CZK
APOSPC49373-1G
APOSPC49373-5G
2-Fluoro-3-iodobenzaldehyde 97%
2-Fluoro-3-iodobenzaldehyde
Vzorec:
C₇H₄FIO MW: 250,009 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15528731 CAS číslo: 146137-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...