6-Fluoro-2-hydroxy-3-nitrobenzotrifluoride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
6-Nitro-α,α,α,3-tetrafluoro-o-cresol, 3-Fluoro-6-nitro-2-(trifluoromethyl)phenol
PC49372-1G
PC49372-250MG
APOSPC49372-1GEA
8270
CZK
APOSPC49372-1G
APOSPC49372-250MG
6-Fluoro-2-hydroxy-3-nitrobenzotrifluoride
6-Fluoro-2-hydroxy-3-nitrobenzotrifluoride
Vzorec:
C₇H₃F₄NO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD24850033 CAS číslo: 1440535-08-9 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...