Tisk…

2,4-Difluoro-3-(trifluoromethyl)benzonitrile 96%

Dodavatel: Apollo Scientific
Varování

Synonyms: α,α,α,2,4-Pentafluoro-m-tolunitrile, 3-Cyano-2,6-difluorobenzotrifluoride

PC49365-1G PC49365-250MG PC49365-5G
APOSPC49365-1GEA 10880 CZK
APOSPC49365-1G APOSPC49365-250MG APOSPC49365-5G
2,4-Difluoro-3-(trifluoromethyl)benzonitrile 96%
2,4-Difluoro-3-(trifluoromethyl)benzonitrile
Vzorec: C₈H₂F₅N
MW: 207,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24850039
CAS číslo: 1440535-21-6
UN: 3276
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR