2-Bromo-6-nitro-4-[(trifluoromethyl)sulphonyl]phenol 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC49361-1G
PC49361-250MG
APOSPC49361-1GEA
7440
CZK
APOSPC49361-1G
APOSPC49361-250MG
2-Bromo-6-nitro-4-[(trifluoromethyl)sulphonyl]phenol 95%
2-Bromo-6-nitro-4-[(trifluoromethyl)sulphonyl]phenol
Vzorec:
C₇H₃BrF₃NO₅S Storage Temperature: Ambient |
MDL Number:
MFCD24850030 CAS číslo: 1440535-14-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...