Tisk…

2,4-Difluoro-3-(trifluoromethyl)styrene

Dodavatel: Apollo Scientific
Varování

PC49345-1G PC49345-250MG
APOSPC49345-1GEA 11380 CZK
APOSPC49345-1G APOSPC49345-250MG
2,4-Difluoro-3-(trifluoromethyl)styrene
2,4-Difluoro-3-(trifluoromethyl)styrene
Vzorec: C₉H₅F₅
Storage Temperature: Refrigerator
MDL Number: MFCD24369700
CAS číslo: 1432075-80-3
UN: 1993
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR